Skip to Content
Merck
All Photos(2)

Key Documents

860348P

Avanti

14:0 PC-d67

Avanti Research - A Croda Brand 860348P, powder

Synonym(s):

1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine-1,1,2,2-d4-N,N,N-trimethyl-d9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H5NO8PD67
CAS Number:
Molecular Weight:
745.35
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860348P-10mg)
pkg of 1 × 25 mg (860348P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860348P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC([2H])([2H])C([2H])([2H])[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H]

InChI

1S/C36H72NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-35(38)42-32-34(33-44-46(40,41)43-31-30-37(3,4)5)45-36(39)29-27-25-23-21-19-17-15-13-11-9-7-2/h34H,6-33H2,1-5H3/t34-/m1/s1/i1D3,2D3,3D3,4D3,5D3,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2,24D2,25D2,26D2,27D2,28D2,29D2,30D2,31D2

InChI key

CITHEXJVPOWHKC-XPIYRYOSSA-N

General description

14:0 PC-d67 is a deuterated phosphocholine (PC) wherein 67 protons of dimyristoyl are replaced by deuterium.
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

14:0 PC-d67 has been used in the preparation of phospholipid bilayer.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860348P-10mg)
5 mL Amber Glass Screw Cap Vial (860348P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Benedetto et al.
The journal of physical chemistry. B, 118(42), 12192-12206 (2014-09-25)
Neutron reflectometry (NR) measurements were carried out to probe the structure and stability of two model biomembranes consisting of 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (POPC) and 1,2-dimyristoyl-sn-glycero-3-phosphatidylcholine (DMPC) phospholipid bilayers hydrated by water solutions of two prototypical room-temperature ionic liquids (RTILs), namely, 1-butyl-3-methyl-imidazolium chloride
Dominique Mias-Lucquin et al.
Journal of structural biology, 209(1), 107411-107411 (2019-11-07)
Dystrophin is a large intracellular protein that prevents sarcolemmal ruptures by providing a mechanical link between the intracellular actin cytoskeleton and the transmembrane dystroglycan complex. Dystrophin deficiency leads to the severe muscle wasting disease Duchenne Muscular Dystrophy and the milder

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service