Skip to Content
Merck
All Photos(2)

Key Documents

840858C

Avanti

16:0-18:2 PA

1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphate (sodium salt), chloroform

Synonym(s):

1-hexadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phosphate (sodium salt); PA(16:0/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H68O8PNa
CAS Number:
Molecular Weight:
694.89
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840858C-25mg)
pkg of 2 × 4 mL (840858C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840858C

concentration

10 mg/mL (840858C-25mg)
25 mg/mL (840858C-200mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C37H69O8P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(39)45-35(34-44-46(40,41)42)33-43-36(38)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h11,13,17-18,35H,3-10,12,14-16,19-34H2,1-2H3,(H2,40,41,42);/q;+1/p-1/b13-11-,18-17-;/t35-;/m1./s1

InChI key

IYNPOFCKGPJAFU-HQFKUYMNSA-M

General description

16:0-18:2 PA or 1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphate is a sodium salt of phosphatidic acid, with substitution of palmitic acid and linolenic acid at the sn-1 and sn-2 positions respectively, in the glycerol backbone.

Application

16:0-18:2 PA or 1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphate (sodium salt) has been used to study the effect of phosphatidic acid (PA) on the cytoplasmic calcium ([Ca2+]cyt) fluctuation in Arabidopsis guard cells by fluorescence imaging.[1] It might be used as a standard in thin-layer chromatography (TLC) for the separation of ginger and grapefruit exosome-like nanoparticles (ELN)-derived lipids.[2] It might also be used in filter-blotting assay, to study the interaction of AT-hook motif-containing nuclear localized protein 4 (Ahl4) with PA.[3]

Packaging

5 mL Clear Glass Sealed Ampule (840858C-200mg)
5 mL Clear Glass Sealed Ampule (840858C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Protocol to Measure the Cytoplasmic Calcium in Arabidopsis Guard Cells
Li L
Bio-protocol, 5(9), e1462-e1462 (2015)
Guangqin Cai et al.
Molecular plant, 13(7), 984-1000 (2020-04-26)
Lipid catabolism in germinating seeds provides energy and substrates for initial seedling growth, but how this process is regulated is not well understood. Here, we show that an AT-hook motif-containing nuclear localized (AHL) protein regulates lipid mobilization and fatty acid
Yun Teng et al.
Cell host & microbe, 24(5), 637-652 (2018-11-20)
The gut microbiota can be altered by dietary interventions to prevent and treat various diseases. However, the mechanisms by which food products modulate commensals remain largely unknown. We demonstrate that plant-derived exosome-like nanoparticles (ELNs) are taken up by the gut

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service