Skip to Content
Merck
All Photos(1)

Key Documents

810509P

Avanti

18:1 Dabsyl PE

Avanti Research - A Croda Brand 810509P, powder

Synonym(s):

1,2-dioleyl-sn-glycero-3-phosphoethanolamine-N-(dabsyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H94N5O10PS
CAS Number:
Molecular Weight:
1048.40
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (810509P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810509P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphoethanolamine is a precursor to phosphatidylcholine and phosphatidylethanolamine. Dabsyl-PE carries contains a dark quencher in the headgroup.

Biochem/physiol Actions

Phosphatidylethanolamine controls membrane fluidity in eukaryotic cells. Phosphoethanolamine (PHOS) has the ability to repress tumour growth both in vitro and in vivo. It possesses anticarcinogenic effects. It can also prevent the mRNA expression of cyclin D1 and vascular endothelial growth factor receptor 1 (VEGFR1). Dabsyl-PE has the ability to quench superfolder green fluorescent protein, (SfGFP).

Packaging

5 mL Amber Glass Screw Cap Vial (810509P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic phosphoethanolamine: the state of the art of scientific production
Anastacio L de B, et al.
Brazilian Journal of Pharmaceutical Sciences , 53(4) (2017)
Gokul Raghunath et al.
Langmuir : the ACS journal of surfaces and colloids, 35(38), 12550-12561 (2019-08-31)
Nickel-chelating lipids offer a convenient platform for reversible immobilization of histidine-tagged proteins to liposome surfaces. This interaction recently found utility as a model system for studying membrane remodeling triggered by protein crowding. Despite its wide array of utility, the molecular
Hyunjin Kim et al.
Journal of lipid research, 54(12), 3531-3538 (2013-09-06)
Membrane-protein interaction plays key roles in a wide variety of biological processes. Although various methods have been employed to measure membrane binding of soluble proteins, a robust high-throughput assay that is universally applicable to all proteins is lacking at present.
Rosie Dawaliby et al.
The Journal of biological chemistry, 291(7), 3658-3667 (2015-12-15)
Adequate membrane fluidity is required for a variety of key cellular processes and in particular for proper function of membrane proteins. In most eukaryotic cells, membrane fluidity is known to be regulated by fatty acid desaturation and cholesterol, although some
Noam Pondé et al.
Ecancermedicalscience, 10, 681-681 (2016-12-03)
The use of unproven forms of therapy in cancer treatment is very common. In Brazil, the distribution by researchers to patients of an investigational agent called phophoethanolamine (PHOS) has led to a widely publicized scientific scandal. PHOS is a precursor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service