Skip to Content
Merck
All Photos(2)

Key Documents

810262P

Avanti

C11 TopFluor Ceramide

N-[11-(dipyrrometheneboron difluoride)undecanoyl]-D-erythro-sphingosine, powder

Synonym(s):

N-[11-(TopFluor)undecanoyl]-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H70N3O3F2
CAS Number:
Molecular Weight:
703.02
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 250 μg (810262P-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810262P

shipped in

dry ice

storage temp.

−20°C

General description

Ceramide, a bioactive sphingolipid belongs to the sphingolipid family of cell lipids. This pore-forming member is insoluble in water. It is a hygroscopic and light sensitive lipid.

Application

C11 TopFluor Ceramide may be used:
  • in the ceramide and B-cell lymphoma-extra-large (Bcl-xL) binding studies to study the effect on ceramide channel formation
  • as a tracer in confocal microscopy to treat T84 cells in order to study its localization in sphingomyelinase (SMase)-induced F-actin structures
  • to study its effect on the migration of pancreatic cancer stem cells (PCSC) using confocal microscopy

Biochem/physiol Actions

Ceramide is known to participate in apoptosis, mediated by mitochondria. Ceramide channels plays a key role in the onset of apoptosis in vivo.

Packaging

5 mL Amber Glass Screw Cap Vial (810262P-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide Channels: Destabilization by Bcl-xL and Role in Apoptosis
Chang KT, et al.
Biochimica et Biophysica Acta, 1848(10 0 0), 2374-2374 (2015)
Microbial sphingomyelinase induces RhoA-mediated reorganization of the apical brush border membrane and is protective against invasion
Saslowsky DE, et al.
Molecular Biology of the Cell, 27(7), 1120-1130 (2016)
Pancreatic ductal adenocarcinoma cell secreted extracellular vesicles containing ceramide-1-phosphate promote pancreatic cancer stem cell motility
Kuc N, et al.
Biochemical Pharmacology, 156, 458-466 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service