610036
Avanti
Syringe Needles (3 pack)
Avanti Research™ - A Croda Brand
Synonym(s):
Extruder Syringe Needle, Replacement needle for syringe
About This Item
Recommended Products
packaging
pkg of 3 ea (needles)
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
Package of 3 needles.
Legal Information
replacement part
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service