Skip to Content
Merck
All Photos(1)

Key Documents

Z765422

Vermiculite 2-3 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131905
NACRES:
NB.35

General description

Raw Vermiculite (sterile), bacteria-and germ-free.

  • High absorbability up to 5 times its own weight
  • Acid and alkali resistant
  • Inorganic
  • Skin-friendly
  • Not electrically conductive
  • Low thermal conduction
  • Sound-absorbing
  • Rot-proof, mold-free, and therefore incorruptible
  • Non-flammable according to DIN 4102 A1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service