Z765198
Tygon® formula E-3603 laboratory tubing
I.D. × O.D. × wall thickness 1/4 in. × 3/8 in. × 1/16 in.
Synonym(s):
DEHP tubing alternative, chemical resistant tubing, laboratory tubing, vacuum tubing
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
Recommended Products
packaging
pkg of 50 feet
manufacturer/tradename
Tygon
I.D. × O.D. × wall thickness
1/4 in. × 3/8 in. × 1/16 in.
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
TYGON® E-3603 non-DEHP laboratory and vacuum tubing delivers the same superior performance you′ve come to expect from TYGON products - but now in a formulation that contains a widely accepted substitute for DEHP plasticizer.
TYGON® E-3603 laboratory tubing is specially formulated for resistance to flex fatigue and abrasion, handles virtually all inorganic chemicals found in the lab, and is less permeable than rubber tubing. The glassy-smooth inner bore helps prevent build-up to facilitate cleaning.
TYGON® E-3603 vacuum tubing shares all these performance features and has extra heavy walls that will withstand a full vacuum at room temperature.
Features/Benefits:
Typical Applications:
TYGON® E-3603 laboratory tubing is specially formulated for resistance to flex fatigue and abrasion, handles virtually all inorganic chemicals found in the lab, and is less permeable than rubber tubing. The glassy-smooth inner bore helps prevent build-up to facilitate cleaning.
TYGON® E-3603 vacuum tubing shares all these performance features and has extra heavy walls that will withstand a full vacuum at room temperature.
Features/Benefits:
- Alternative to DEHP tubing
- New formulation delivers the same excellent performance
- Contains no BPA
- Outstanding chemical resistance
- Lot-to-lot consistency for reproducible results
- Increases productivity in peristaltic pumps - outlasts other clear tubing 2 to 1
- Smooth, polished inner bore
- Non-oxidizing and non-contaminating
- Slips easily over fittings and grips securely
- Coils marked at 1-foot intervals for easy measuring
- Meets FDA criteria
Typical Applications:
- General laboratory
- Analytical instruments
- Peristaltic and vacuum pumps
- Ideal for condensers, incubators, desiccators, gas lines and drain lines
Legal Information
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service