Skip to Content
Merck
All Photos(1)

Key Documents

Z742592

Accuris SmartDoc

2.0 Imaging System with blue light illumination base, AC/DC input 230 V AC, EuroPlug

Synonym(s):

gel documentation equipment, transilluminator system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106000
NACRES:
NB.24

Pricing and availability is not currently available.

feature

12V DC power supply included
High Intensity Blue LED; Orange PMMA photo filter and view cover
includes gel prep/cutting platform, blue light illumination base, orange filter cover, imaging enclosure and orange photo filter

packaging

pkg of 1

manufacturer/tradename

Benchmark Scientific E5001-SDB-E        

W × D × H

23 cm × 19 cm × 22 cm, 2.6lbs (1.2kg) weight

compatibility

smartphones and tablets with camera

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
P6354P4013P9511
assay

≥97% (TLC), ≥99% (GC)

assay

≥99% (TLC)

assay

≥99%

assay

≥98%

functional group

phospholipid

functional group

phospholipid

functional group

phospholipid

functional group

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

powder

form

lyophilized powder

form

powder

form

powder

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

-

General description

Introducing the SmartDoc 2.0. This complete system has been developed for safely viewing nucleic acids in agarose gels and also for capturing quality digital images with a smart phone camera. Blue light is used instead of UV, which is safer for lab personnel and will not damage DNA samples. The illumination base emits an intense blue light with a peak output of 460nm for optimum excitation of nucleic acids stained with safe green dyes (SYBR Green, SmartGlow, GelStar, GelGreen, etc.) The orange cover fits on top of the illumination base and acts as a filter to absorb blue light, allowing visualization of the fluorescing samples. The cover can also be set at an angle for gel access and cutting out DNA bands for further study. The SmartDoc imaging enclosure includes an orange photo filter for photography of the gels with a smartphone. An extending adapter can be inserted for full dimension imaging of gels up to 15X15 cm or removed for close up pictures. The enclosure can also be used directly on any other existing UV or blue light trans illuminator. Additional filter are available for imaging gels on a UV trans illuminator.

Features:
  • Ideal for common safe stains (SmartGlow, SYBR Green, GelGreen, etc)
  • Visible blue light will not damage DNA
  • Quality gel imaging with any smart phone
  • Gel access for DNA band cutting
  • Imaging enclosure can also be used on existing trans illuminators
  • Maximum gel size: 15X15 cm

Legal Information

Accuris is a trademark of Benchmark Scientific, Inc.
SmartDoc is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service