Z742569
PlateFuge™ Microplate Microcentrifuge with rotor and plate carrier
AC/DC input 230 V AC, UK plug
Synonym(s):
Microplate centrifuge, Plate rotor centrifuge
About This Item
Recommended Products
feature
CE compliant
to hold 2 PCR plates
to hold 24 microtiter plates
4 S Deceleration:
packaging
pkg of 1
manufacturer/tradename
Benchmark Scientific C2000-E-UK
parameter
2550 rpm
600 × g
technique(s)
centrifugation: suitable
W × D × H
9.2 in. (23 cm) × 10.2 in. (26 cm) × 7.75 in. (19.7 cm)
weight
9 lb (4 kg)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service