Z676470
Double-Safe™ PTFE coated thermometer
L 305 mm, temperature -20-150 °C, immersion level, total
All Photos(2)
About This Item
UNSPSC Code:
41112303
NACRES:
NB.79
Recommended Products
feature
immersion level total
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B60400-1800
greener alternative product characteristics
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
temperature measuring range
-20-150 °C
L
305 mm
greener alternative category
, Aligned
Related Categories
General description
Double-Safe thermometers are PTFE coated Easy-Read thermometers with black Enviro-Safe liquid and yellow back lead free glass. Double-Safe thermometers prevent contamination of the glass and liquid in the event of breakage. Double-Safe is the ideal replacement for mercury thermometers banned (or partially banned) in many US states and most European countries.
- PTFE coating contains liquid and glass in case of breakage
- Non-toxic, non-hazardous, biodegradable black Enviro-Safe liquid
- Available with statement of accuracy traceable to NIST and DKD
- Lead-free glass and recyclable packaging
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical without mercury. Click here for more information.
Legal Information
Double-Safe is a trademark of H-B Instrument Co.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)