Z609706
Bohlender™ PTFE tubing
I.D. × O.D. 0.5 mm × 1.0 mm, L 10 m
Synonym(s):
PTFE Tubing
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 10 m
manufacturer/tradename
Bohlender
availability
not available in USA and Canada
I.D. × O.D.
0.5 mm × 1.0 mm
L
10 m
wall size
0.25 mm
Looking for similar products? Visit Product Comparison Guide
Legal Information
Bohlender is a trademark of Bohlender GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service