Z561495
Thermco® precision ground-glass joint thermometer
temperature -10-250 °C, immersion L 50 mm
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Joint
joint ST/NS10/30
description
spirit-filled
feature
subdivision 1.0 °C
manufacturer/tradename
Thermco Products, Inc.
temperature measuring range
-10-250 °C
L
350 mm
Looking for similar products? Visit Product Comparison Guide
Legal Information
Thermco is a registered trademark of Thermco Products, Inc.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service