Z543748
Aldrich® thermometer adapter for Aldrich student glass kit
joint: ST/NS 14/20, pkg of 1 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Recommended Products
General description
This Aldrich thermometer adapter has a standard 14/20 ground-glass joint which provides a high-quality, low-cost solution for school laboratories and research facilities. Contact our Glass Center to discuss your custom kit requirements at [email protected]
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service