Z530433
Aldrich® Claisen distillation head
Joints: ST/NS 14/20 (3), thermometer joint: ST/NS 10/18
All Photos(1)
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Recommended Products
Joint
thermometer joint: ST/NS10/18
Joints: ST/NS14/20 (3)
immersion depth
50 mm
Related Categories
General description
Claisen adapter with a thermometer joint. Male side-arm drip-tip joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)