Z516007
Schlenk reaction and storage tube
female joint: ST/NS 14/20, capacity 50 mL
Synonym(s):
storage tube, tube flask, tube storage
About This Item
Recommended Products
material
colorless cap
colorless tube
round bottom
Joint
female joint: ST/NS14/20
feature
graduations: no
capacity
50 mL
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | A9935 | A6907 | A9794 |
---|---|---|---|
affinity isolated antibody | affinity isolated antibody | affinity isolated antibody | affinity isolated antibody |
Ouchterlony double diffusion: suitable, quantitative precipitin assay: suitable | Ouchterlony double diffusion: suitable, immunoelectrophoresis: suitable | direct ELISA: 1:10,000 | direct ELISA: 1:50,000, dot blot: 1:60,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200, western blot (chemiluminescent): 1:60,000 |
goat | goat | goat | goat |
2-8°C | 2-8°C | −20°C | 2-8°C |
lyophilized powder | PBS suspension | buffered aqueous solution | buffered aqueous solution |
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service