Z341517
Hirschmann® Ceramus® bottle-top dispenser, adjustable volume
dispensing volume 10.0-60.0 mL, precision: 0.1 CV%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
subdivision 1.0 mL
with recirculating valve
manufacturer/tradename
Hirschmann 9362000
dispensing volume measuring range
10.0-60.0 mL, precision: 0.1 CV%
volume
10-60 mL
fitting
(A 28)
(A45)
S 40
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Chemically inert − unique ceramic dispenser piston in a Duran® borosilicate glass barrel
- Micro-step volume adjustment − ensures reproducibility
- Rotates on the bottle − for convenience
- Recirculation system − for fast priming without loss of reagent
- Lifetime calibration − supplied with individual serial number and Test Certificate
Includes adapters for:
- GL 45 threads/Pyrex® and Duran® media bottles
- GL 32 threads/Aldrich Sealed for Quality and Sure/Seal bottles
- GL 38 threads/Gallon bottles
- S40 threads
Specifications:
- Piston − 99.7% pure Al2O3
- Barrel − Duran® borosilicate glass
- Valve block and housing − ECTFE
- Valves − borosilicate glass ball and seat (HF: ceramic); Hastelloy® discharge valve spring
- Suction and discharge tubes − FEP
- Sterilization − Steam sterilizable at 121°C/2 bar, complete unit without disassembly
- Luer-lock connection − for filters and drying tubes
Legal Information
Ceramus is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
DURAN is a registered trademark of DWK Life Sciences
Hastelloy is a registered trademark of Haynes International, Inc.
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service