Key Documents
Z290882
Sigma-Aldrich
PEEK Tubing Cutter
Synonym(s):
PEEK Tubing Cutter
About This Item
UNSPSC Code:
41115700
eCl@ss:
32035301
NACRES:
SB.52
Pricing and availability is not currently available.
General description
Makes burr-free, perpendicular cuts through polymer tubing with outside diameters from 1/16 in. to 1/8 in.. Designed for cutting PEEK tubing, but also easily slices through PTFE and Tefzel capillary tubing. Compact design includes a safety locking mechanism. One spare blade included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)