Key Documents
Z278319
Conical reaction vial
capacity 8 mL
About This Item
UNSPSC Code:
41121700
NACRES:
NB.43
material
colorless tube
glass tube
screw top neck
sterility
non-sterile
feature
graduations: no
External thread
packaging
pack of 1 ea
manufacturer/tradename
Ace Glass D104154
capacity
8 mL
General description
Thin-wall with threaded outer 14/10 joint, cap with hole, PTFE-faced silicone rubber septum, and FETFE® O-ring.
The product does NOT have the side arm opening. It is the product shown on the left of the photo.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)