Skip to Content
Merck
All Photos(2)

Key Documents

Z272078

NMR tube caps

NMR tube size 3 mm

Synonym(s):

cap tubes, nmr tubes, plastic end cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
SB.32

packaging

pkg of 100 ea

manufacturer/tradename

Wilmad 521-P-100

NMR tube size

3 mm

color

red

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service