Z267732
Round crystal window
NaCl, diam. × thickness 25 mm × 5 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
NaCl
technique(s)
IR spectroscopy: suitable
diam. × thickness
25 mm × 5 mm
transmittance
40,000-62,500 (wavenumbers (cm-1))
refractive index
n/D 1.52 (at 2 μm)
Looking for similar products? Visit Product Comparison Guide
General description
Very soft, water soluble crystal; low cost and good transmission range; fogs.
Cut and polished quality crystal windows.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service