Key Documents
Z266086
Sigma-Aldrich
TLC desiccating cabinet
provides complete visability of stored TLC plates under desiccating conditions
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
General description
Acrylic cabinet provides complete visability of stored TLC plates under desiccating conditions. Stores up to 24 (20 × 20 cm) plates. Plates should be oven dried before storage for optimal results.
Choose from one of the most recent versions:
Certificates of Analysis (COA)