Z261289
Stainless steel 316 syringe needle, noncoring point
gauge 20, L 6 in.
Synonym(s):
316 Stainless Needle, Noncoring Needle
All Photos(1)
About This Item
Recommended Products
description
Micro-Mate Hub with 316 Stainless Steel Tubing
packaging
pack of 1 ea
manufacturer/tradename
Popper 9863
L
6 in.
gauge
20
General description
Specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. Female Luer hubs and needles are made of 316 SS. Noncontaminating, easy to clean and resistant to corrosion.
Micro-Mate Hub with 316 Stainless Steel Tubing
Micro-Mate Hub with 316 Stainless Steel Tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)