Z258709
Ace vacuum-sublimation apparatus
Complete apparatus, H × I.D. 200 mm × 60 mm
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.76
manufacturer/tradename
Ace Glass 802340
H × I.D.
200 mm × 60 mm
General description
The Ace vacuum-sublimation apparatus features a greaseless O-ring sealed flange with a quick-release clamp. The top flange is ground flat, bottom mating flange has a groove for a CAPFE (fluororesin encapsulated silicone) O-ring. The complete apparatus includes an outer body, inner top, CAPFE O-ring, and quick-release clamp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)