Z231622
Duran® watch glasses
O.D. 125 mm
Synonym(s):
213215208
Select a Size
R$4,567.00
Availability
Estimated to ship onMarch 10, 2025
Select a Size
About This Item
R$4,567.00
Availability
Estimated to ship onMarch 10, 2025
manufacturer/tradename
DURAN®
O.D.
125 mm
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | HPA014715 | HPA035097 | HPA067561 |
---|---|---|---|
rabbit | rabbit | rabbit | rabbit |
affinity isolated antibody | affinity isolated antibody | affinity isolated antibody | affinity isolated antibody |
unconjugated | unconjugated | unconjugated | unconjugated |
Prestige Antibodies® Powered by Atlas Antibodies | Prestige Antibodies® Powered by Atlas Antibodies | Prestige Antibodies® Powered by Atlas Antibodies | Prestige Antibodies® Powered by Atlas Antibodies |
buffered aqueous glycerol solution | buffered aqueous glycerol solution | buffered aqueous glycerol solution | buffered aqueous glycerol solution |
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service