Z231517
Duroplan petri dishes
O.D. × H 80 mm × 20 mm
Synonym(s):
217554307
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
sterility
non-sterile
feature
lid
packaging
case of 10 ea
manufacturer/tradename
DURAN® 217554307
availability
not available in USA and Canada
O.D. × H
80 mm × 20 mm
dish capacity
~100 mL
dish diam.
80 mm
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Supplied in pairs as dish and cover. These petri dishes are manufactured to give even distribution of culture media and distortion-free transparency. Meet DIN 12339 standard.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service