Skip to Content
Merck
All Photos(1)

Key Documents

Z222984

Sigma-Aldrich

Stainless steel desiccator cabinet

size small

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104510
NACRES:
NB.43

size

small

W × L × H

31.8 cm × 31.1 cm × 31.1 cm

chamber W × L × H

29.2 cm × 25.4 cm × 30.5 cm

Looking for similar products? Visit Product Comparison Guide

General description

Polished finish, stackable with double-action bar lock; two sides and door of 3/16 in. glass. Shelf size 24.8 x 22.9 cm
Type 302 SS with glass door and walls and two expanded SS shelves with 1/2 in. adjustment. One-piece neoprene gasket seal and a full-length door hinge for an air-tight, moisture-proof closure. Shelves are adjustable and come in 2 different sizes. Manual pressure/vacuum relief valve and SS desiccant tray. No vacuum or pressure should be applied.


Applications:
  • Storing reagents, IR cells or soil samples
  • Drying biological specimens
  • Cooling crucibles
  • Storage sludge samples

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service