Z212644
Ace powder dispensing funnel
capacity 100 mL
Synonym(s):
funnel powder, glass funnel, glass funnels
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
Recommended Products
Joint
joint ST/NS24/40
manufacturer/tradename
Ace Glass 723930
capacity
100 mL
Related Categories
General description
For powders and solids up to 25 mesh. Features a flexible PTFE screw thread wrapped on a precision PTFE-ceramic shaft contained within a precision bore housing. Completely inert materials allow flushing with solvents. No.15 Ace-Thred affords easy disassembly and cleaning.
Note: This funnel operates on a screw feed principal. The rate or amount of powder added is controlled by turning the knob which feeds the powder into the funnel like a conveyor.
Note: This funnel operates on a screw feed principal. The rate or amount of powder added is controlled by turning the knob which feeds the powder into the funnel like a conveyor.
Choose from one of the most recent versions:
Certificates of Analysis (COA)