Z205389
Eye-protection holder
clear acrylic, H × W × D 3 1/4 in. × 9 in. × 6 3/4 in.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear acrylic
feature
lid: no
Holds safety glasses
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F24877-0001
H × W × D
3 1/4 in. × 9 in. × 6 3/4 in.
back plate size
6 in.
General description
Sturdy holder provides workers with easy access to safety glasses or goggles. Wall mountable or freestanding.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service