Skip to Content
Merck
All Photos(1)

Key Documents

Z154903

Sigma-Aldrich

Disposable polystyrene weighing dishes

L × W × H 135 mm × 135 mm × 20 mm, white

Synonym(s):

Disposable Weighing Dishes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.71

material

polystyrene
rectangular

packaging

pack of 100 ea
case of 500 ea

manufacturer/tradename

Sigma-Aldrich

L × W × H

135 mm × 135 mm × 20 mm

dish L × W

135 mm × 135 mm

color

white

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service