Recommended Products
Joint
joint: ST/NS24/40
General description
Provides a ST/NS: joint connection to single bulb ball-tube flasks and a hose connector to motor drive.
Choose from one of the most recent versions:
Certificates of Analysis (COA)