Z107557
Aldrich® low form dewar flask
capacity 220 mL, cylindrical, depth 6 1/2 cm
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
material
borosilicate glass
Joint
Joints neck (65)
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
I.D.
6.5 cm
capacity
220 mL
depth
6 1/2 cm
Looking for similar products? Visit Product Comparison Guide
General description
Aldrich low form dewar flask can be used in reactions requiring low temperatures and magnetic stirring. It has a shallow design in hemispherical or cylindrical (semi-flat bottom) configurations. The dewar flask is made of borosilicate glass with protective outer aluminum housing.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service