WHA77073500
Whatman® 850-DS 8 channel filter plates
0.2 μm, Nylon membrane, non-sterile, 50 ea
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NC.01
Recommended Products
Product Name
Whatman® 850-DS 8-channel filter plate, Nylon membrane, pore size 0.2 μm, pack of 50 ea
material
Nylon membrane
packaging
pack of 50 ea
manufacturer/tradename
Whatman 7707-3500
pore size
0.2 μm pore size
binding type
non-treated surface
Related Categories
General description
Save time and eliminate errors associated with manual sampling. Use Whatman 850-DS 8-channel filter plates in your Agilent 850-DS Dissolution Sampling Station
Application
The filter plates are exclusively designed for use with the optional filter module on the Agilent 850-DS Dissolution Sampling Station to simplify filter replacement between timepoints. Reliable alignment of the liquid path increases productivity in two ways. One, by reducing the risk of jamming, and two, by reducing leaks that may occur with manual sampling or other dissolution sample preparation systems.
Features and Benefits
- Automated processing: up to 8 samples simultaneously
- Readily available: in a wide range of pore sizes and materials
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)