WHA68962502
Whatman® GD/X syringe filters
polyethersulfone membrane, pore size 0.2 μm, diam. 25 mm, sterile, pk of 50 ea
Synonym(s):
Whatman filter, Z286923, syringe filter, syringe filters
About This Item
Recommended Products
material
polyethersulfone membrane
polypropylene housing
sterility
sterile
feature
glass fiber prefilter
packaging
pk of 50 ea
manufacturer/tradename
Whatman 6896-2502
Whatman Article No. 28420548 (US reference)
diam.
25 mm
volume
≤25 mL
pore size
0.2 μm
fitting
female Luer inlet (lock)
male Luer outlet
Related Categories
General description
Features and Benefits
- Reduces need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters
- Female luer lock for leak-free connections
- High-efficiency Whatman GMF 150 and GF/F pre-filter media
- Eliminates or greatly reduces the need to carry out separate prefiltration step for difficult-to-filter samples
Suitability
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)