Skip to Content
Merck
All Photos(1)

Key Documents

W326801

Sigma-Aldrich

3,4-Dimethyl-1,2-cyclopentadione

≥97%, FG

Synonym(s):

Caramel pentadione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O2
CAS Number:
Molecular Weight:
126.15
FEMA Number:
3268
EC Number:
Council of Europe no.:
2234
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.075
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥97%

mp

68-72 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caramel; maple; sweet

storage temp.

2-8°C

SMILES string

CC1CC(=O)C(=O)C1C

InChI

1S/C7H10O2/c1-4-3-6(8)7(9)5(4)2/h4-5H,3H2,1-2H3

InChI key

WGAVDEVFJDQIMZ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service