W320900
5-Methyl-2-thiophenecarboxaldehyde
FG
Synonym(s):
bread thiophene, NSC 87542
About This Item
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Fragrance grade
Halal
Kosher
Agency
follows IFRA guidelines
meets purity specifications of JECFA
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
refractive index
n20/D 1.583 (lit.)
bp
114 °C/25 mmHg (lit.)
density
1.17 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
Organoleptic
almond; bread; cherry; sweet
storage temp.
2-8°C
SMILES string
[H]C(=O)c1ccc(C)s1
InChI
1S/C6H6OS/c1-5-2-3-6(4-7)8-5/h2-4H,1H3
InChI key
VAUMDUIUEPIGHM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
172.4 °F - closed cup
Flash Point(C)
78 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service