TEM-LC400CUUL
Ultrathin Lacey Carbon Supported Copper Grid
size 400 mesh, box of 25
Synonym(s):
LACEY CARBON FILM CU #400 ULTRA THIN 25/BX
All Photos(1)
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Recommended Products
packaging
box of 25
size
400 mesh
Related Categories
General description
Ultrathin Lacey Carbon (4-6 nm) Supported Grid: Copper Grid, 400 mesh. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)