SYNV381512
Synthware™ vacuum trap with #15 o-ring joint and high vacuum valve
body O.D. 57 mm, body L 300 mm
Synonym(s):
Synthware V381512
All Photos(1)
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
Recommended Products
manufacturer/tradename
V381512
body L
300 mm
body O.D.
57 mm
Related Categories
General description
Synthware™ vacuum trap features a two-piece design with a 0-12 mm high vacuum valve as the shut-off. Connection is made via a 57 mm O-ring flat flange and a #331 Viton® O-ring.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)