SYNS391656
Synthware™ high vacuum double oblique glass stopcock
bore 4 mm, stem O.D. 10 mm, Plug Size: 16.2/56
Synonym(s):
Synthware S391656
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
S391656
bore
4 mm
stem O.D.
10 mm
Related Categories
General description
Synthware™ glass stopcock features solid glass plug. Barrel and plug are precision ground with corresponding numbers on the barrel and plug. With constant pressure retention spring and clip.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)