SYNF271018M
Synthware™ fritted gas dispersion tube with 15 degree angle
disc diam. 10 mm, porosity: medium, stem O.D. × L 8 mm × 150 mm
Synonym(s):
Synthware F271018M, glass gas dispersion tube
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
material
colorless tube
glass tube
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F271018M
disc diam.
10 mm
stem O.D. × L
8 mm × 150 mm
Related Categories
General description
The 15° angle permits tube to be inserted into vessels where a large opening is not available. Stem is 8 mm O.D. × 150 mm length, XC stands for extra coarse porosity.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)