SYNF21342L
Synthware™ four-neck round-bottom flask with vertical side necks
capacity 2,000 mL, center joint: ST/NS 34/45, side joint: ST/NS 24/40
Synonym(s):
Synthware F21342L, 4-neck reaction flask
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Recommended Products
material
borosilicate glass flask
four-neck
round bottom
Joint
center joint: ST/NS34/45
side joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F21342L
capacity
2,000 mL
Related Categories
General description
With identical vertical side joints.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)