SYNC579110
Synthware™ water jacketed distillation column with removable hose connections
joint: ST/NS 19/22, jacket H 110 mm
Synonym(s):
Synthware C579110, glass distilling column
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C579110
jacket H
110 mm
Related Categories
General description
Indentations at the bottom for supporting packing material. Jacket permits the column to be used as a condenser.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)