SYNA761420
Synthware™ flushing adapter with T-bore PTFE stopcock
joint: ST/NS 14/20
Synonym(s):
Synthware A761420, glass flushing adapter
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A761420
Related Categories
General description
With 2 mm T-bore PTFE stopcock.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)