Recommended Products
Assay
98%
form
powder
shelf life
3 years under the recommended conditions
color
white
mp
181-187 °C
SMILES string
OP(O)(=O)CCCCCCCCCCCCP(O)(O)=O
InChI
1S/C12H28O6P2/c13-19(14,15)11-9-7-5-3-1-2-4-6-8-10-12-20(16,17)18/h1-12H2,(H2,13,14,15)(H2,16,17,18)
InChI key
BEPFDRNIALBIKQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service