All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C85H153F4N5O33
Molecular Weight:
1849.13
UNSPSC Code:
51171641
NACRES:
NA.22
Recommended Products
Assay
>90%
form
solid or viscous liquid
reaction suitability
reagent type: chelator
reagent type: cross-linking reagent
reagent type: fluorescent-labelling reagent
polymer architecture
shape: linear
functionality: monofunctional
shipped in
ambient
storage temp.
−20°C
Related Categories
Features and Benefits
DOTA-tris(TBE)-amido-dPEG ®₂₄-TFP ester contains a DOTA-tris(TBE) metal chelator for radio-labelling connected through a hydrophilic dPEG spacer to a TFP ester which can react with a primary amine.
Legal Information
Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service