Skip to Content
Merck
All Photos(1)

Key Documents

QBD10954

Sigma-Aldrich

Bis-dPEG®13-NHS ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H64N2O21
CAS Number:
Molecular Weight:
884.92
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

polymer architecture

shape: linear
functionality: homobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(ON1C(CCC1=O)=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(ON2C(CCC2=O)=O)=O

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
QBD10246QBD10127QBD11086
form

solid or viscous liquid

form

solid or viscous liquid

form

solid or viscous liquid

form

solid or viscous liquid

assay

>90%

assay

>90%

assay

>90%

assay

>90%

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

reaction suitability

reagent type: chemical modification reagent, reagent type: cross-linking reagent
reactivity: amine reactive

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

Features and Benefits

Bis-dPEG®13-NHS ester is a homobifunctional dPEG® crosslinking product which allows conjugation to two different moieties, each containing free amines. The single molecular weight dPEG® spacer (50.1 Å) provides precise distance control between conjugated moieties. N-hydroxysuccinimide (NHS) ester versions are highly reactive to free amines.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service