MT51344022
Mettler Toledo InLab® 73 series conductivity probe
InLab® 731, cable L 2 m, Mini-DIN connector
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
epoxy shaft
description
4 pole graphite cell
cell constant 0.57 cm-1, epoxy shaft
feature
Mini-DIN connector
manufacturer/tradename
Mettler-Toledo 51344022
L × diam.
120 mm × 12 mm
cable L
2 m
General description
InLab® 73 series have 4-electrode (4 pole) carbon sensors, ideal for general purpose use.
Supplied with quality certificate, integrated temperature sensor, plug and cable.
Intended use and principles
These products combine a state-of-the-art conductivity cell with a thermistor which is integrated in the sensor body enabling automatic temperature compensating (ATC) to
provide an easy to use, reliable and accurate sensor. The electrode design minimizes the potential for external interferences from sample containers or other sensors. These
sensors are equipped with cables in various lengths and two types of plugs to match Mettler TOLEDO conductivity meters, with and without Intelligent Sensor Management (ISM).
Intelligent Sensor Management (ISM®)
The Mettler TOLEDO dual channel portable meters SevenGo Duo (pro) SG23, 68 and 78 have the ability to automatically recognize ISM sensors upon connection, transfer calibration data to and from the sensor etc. For more information regarding ISM® please read the respective chapter of the operating instructions supplied with the meter.
Supplied with quality certificate, integrated temperature sensor, plug and cable.
Intended use and principles
These products combine a state-of-the-art conductivity cell with a thermistor which is integrated in the sensor body enabling automatic temperature compensating (ATC) to
provide an easy to use, reliable and accurate sensor. The electrode design minimizes the potential for external interferences from sample containers or other sensors. These
sensors are equipped with cables in various lengths and two types of plugs to match Mettler TOLEDO conductivity meters, with and without Intelligent Sensor Management (ISM).
Intelligent Sensor Management (ISM®)
The Mettler TOLEDO dual channel portable meters SevenGo Duo (pro) SG23, 68 and 78 have the ability to automatically recognize ISM sensors upon connection, transfer calibration data to and from the sensor etc. For more information regarding ISM® please read the respective chapter of the operating instructions supplied with the meter.
Other Notes
The InLab® 731 probe is not protected against ingress of water and dust (IP67 rated).
Legal Information
ISM is a registered trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service