Key Documents
MT51343174
Mettler Toledo InLab® Cool pH electrode
for use in low temperature samples
About This Item
UNSPSC Code:
41115604
NACRES:
NB.79
material
glass (LoT)
glass membrane (polymer)
description
S7 connector
without cable
manufacturer/tradename
Mettler-Toledo 51343174
L × W
120 mm × 12 mm
General description
The Mettler Toledo InLab® Cool is a glass-body, combination pH electrode with S7 screw head. Reference electrode is filled with Friscolyte-B electrolyte for low temperature samples.
By filling the reference electrode with Friscolyte-B® electrolyte the electrode can be used in samples down to a temperature of minus thirty degrees Centigrade.
ARGENTHAL reference system improves service life and measurement accuracy at differing sample temperatures.
Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
  • ARGENTHAL reference
  • Friscolyte-B electrolyte
  • Detachable cable
Legal Information
ARGENTHAL is a trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)