M26615
4′-Methylacetophenone
95%
Synonym(s):
Methyl p-tolyl ketone
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3C6H4COCH3
CAS Number:
Molecular Weight:
134.18
Beilstein:
606053
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
refractive index
n20/D 1.533 (lit.)
bp
226 °C (lit.)
mp
22-24 °C (lit.)
density
1.004 g/mL at 20 °C
1.005 g/mL at 25 °C (lit.)
SMILES string
CC(=O)c1ccc(C)cc1
InChI
1S/C9H10O/c1-7-3-5-9(6-4-7)8(2)10/h3-6H,1-2H3
InChI key
GNKZMNRKLCTJAY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
179.6 °F - closed cup
Flash Point(C)
82 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Koji Uwai et al.
Bioorganic & medicinal chemistry, 16(3), 1084-1089 (2007-11-17)
Stereoselective reductive metabolism of various p-substituted acetophenone derivatives was studied using isolated rat liver 3alpha-hydroxysteroid dehydrogenase (3alpha-HSD). Kinetic experiments were performed and analyzed by measuring the products by HPLC using a chiral column. The results demonstrated that the presence of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service