Skip to Content
Merck
All Photos(2)

Key Documents

I5634

Sigma-Aldrich

4-(p-Iodophenyl)butyric acid

≥98%

Synonym(s):

4-Iodobenzenebutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11IO2
CAS Number:
Molecular Weight:
290.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

storage temp.

2-8°C

SMILES string

OC(=O)CCCc1ccc(I)cc1

InChI

1S/C10H11IO2/c11-9-6-4-8(5-7-9)2-1-3-10(12)13/h4-7H,1-3H2,(H,12,13)

InChI key

OGOMLUBUDYFIOG-UHFFFAOYSA-N

Application

Reactant involved in:
  • Synthesis of meta- and paracyclophanes containing unsaturated amino acids
  • Intramolecular Friedel-Crafts reactions for synthesis of 1-tetralones

Involved in investigations of pharmacological activity pertaining to:
  • Apoptosis, proliferation, and histon deacetylase activity in human colorectal cancer cells
  • Portable albumin binders for extending the half-life of contrast agents

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service