Skip to Content
Merck
All Photos(2)

Key Documents

GF62374765

Iron

foil, 25mm disks, thickness 0.075mm, hard, 99.5%

Synonym(s):

Iron, FE000290

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.23

Pricing and availability is not currently available.

Assay

99.5%

form

foil

manufacturer/tradename

Goodfellow 623-747-65

resistivity

9.71 μΩ-cm

diam. × thickness

25 mm × 0.075 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Jun-Won Jang et al.
    Water science and technology : a journal of the International Association on Water Pollution Research, 59(12), 2503-2507 (2009-06-23)
    Zero valent iron has been successfully used for the degradation of a wide range of contaminants. However, this reaction of using ZVI particle produces a large quantity of iron sludge. To solve the problem, we report the synthesis of self-organized
    Arnold L Demain et al.
    Applied microbiology and biotechnology, 73(1), 55-59 (2006-04-20)
    When tetanus toxin is made by fermentation with Clostridium tetani, the traditional source of iron is an insoluble preparation called reduced iron powder. This material removes oxygen from the system by forming FeO(2) (rust). When inoculated in a newly developed
    Maria Wilhelm et al.
    Clinical nephrology, 81(4), 251-258 (2014-03-25)
    Hyperphosphatemia in advanced chronic kidney disease (CKD) necessitates the use of phosphate binders. This in vitro study assessed phosphate binding and Fe release properties of the novel iron-based phosphate binder PA21. Phosphate adsorption and Fe release were assessed under conditions
    Antonino Missineo et al.
    Infection and immunity, 82(6), 2448-2459 (2014-04-02)
    Staphylococcus lugdunensis is a coagulase-negative staphylococcus that is a commensal of humans and an opportunistic pathogen. It can cause a spectrum of infections, including those that are associated with the ability to form biofilm, such as occurs with endocarditis or
    Catherine F M Bowden et al.
    Biochemistry, 53(14), 2286-2294 (2014-03-22)
    The Isd (iron-regulated surface determinant) system is a multiprotein transporter that allows bacterium Staphylococcus aureus to take up iron from hemoglobin (Hb) during human infection. In this system, IsdB is a cell wall-anchored surface protein that contains two near iron

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service